Anti-CD3 Mouse Monoclonal Antibody [clone: OKT3] (FITC)
Leverandør: STEMCELL Technologies
100-0285 100-0286
STMC100-0285EA 0 NOK
STMC100-0285 STMC100-0286
Anti-CD3 Mouse Monoclonal Antibody [clone: OKT3] (FITC)
Antistoffer
Mouse monoclonal IgG2a, antibody against human CD3, FITC-conjugated.
  • T cells
The OKT3 antibody reacts with an extracellular conformational epitope on the ~20 kDa CD3ε subunit of the human T cell receptor (TCR)/CD3 complex, which is expressed on the surface of ~95% of mature T cells and NKT cells, and variably on thymocytes. A majority of T cell neoplasms also express CD3. CD3 is a dimer assembled from combinations of CD3γ, δ, ε, η, and ζ subunits, which are structurally related type 1 transmembrane proteins and members of the immunoglobulinMore Product Information