Anti-CD45R (B220) Rat Monoclonal Antibody [clone: RA3-6B2] (FITC)
Leverandør: STEMCELL Technologies
60019FI 100-0423
STMC60019FIEA 0 NOK
STMC60019FI STMC100-0423
Anti-CD45R (B220) Rat Monoclonal Antibody [clone: RA3-6B2] (FITC)
Antistoffer
Rat monoclonal IgG2a antibody against human, mouse, cat CD45R (B220).
  • B cells
  • Leukocytes
  • Lymphocytes
The RA3-6B2 antibody reacts with an epitope on the extracellular domain of the exon A‐restricted isoform of mouse CD45. CD45 is a member of the protein tyrosine phosphatase family and functions in a number of immunoregulatory processes, including cell activation, growth, differentiation, and oncogenic transformation. The large cytoplasmic portion of CD45 contains two tyrosine phosphatase domains (one of which is enzymatically active) that are involvedMore Product Information