Anti-CD11b Rat Monoclonal Antibody [clone: M1/70] (Biotin)

Leverandør: STEMCELL Technologies
60001BT 100-0439 100-0438
STMC60001BTEA 0 NOK
STMC60001BT STMC100-0439 STMC100-0438
Anti-CD11b Rat Monoclonal Antibody [clone: M1/70] (Biotin)
Antistoffer
Rat monoclonal IgG2b antibody against human, mouse, rhesus CD11b, biotin-conjugated.

  • Granulocytes
  • Subsets
  • Monocytes

The M1/70 antibody reacts with CD11b, an ~170 kDa type 1 transmembrane glycoprotein which associates non-covalently with CD18 to form the heterodimeric Mac-1 receptor. Through its interactions with ligands such as ICAM-1 (CD54), ICAM-2 (CD102), ICAM-4 (CD242), iC3b, and fibrinogen, Mac-1 functions in several processes, including the adherence of neutrophils and monocytes to stimulated endothelium and phagocytosis of complement-coatedMore Product Information
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn CD11b
Klonalitet Monoclonal
Konjugering Biotin
Klon M1/70
Reaktivitet Baboon,Rabbit,Rhesus monkey,Chimpanzee,Human
Vert Rat
Gen-ID 16409
Isotype IgG2b kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR