ImmunoCult™ Human B Cell Expansion Kit
Leverandør: STEMCELL Technologies
100-0645
STMC100-0645EA 0 NOK
STMC100-0645
ImmunoCult™ Human B Cell Expansion Kit
Media, Cellekultur
Serum-free culture kit for expansion of human B cells.
Reliably expand human B cells that are ready for use in downstream assays and applications - without the use of serum, specialized cultureware, or problematic feeder cells. ImmunoCult™ Human B Cell Expansion Kit comprises ImmunoCult™-ACF Human B Cell Expansion Supplement and ImmunoCult™-XF B Cell Base Medium, which together ensure consistent activation and expansion of human B cells and their maturation to plasma cells. B cells can be harvested and used directly in downstream applications after just one week or less in culture.
For convenience, all kit components are also available for individual purchase. This kit is compatible with many other upstream and downstream products. For example, use EasySep™ cell separation kits to isolate a range of B cell subsets - such as pan-B cells, memory B cells, and naïve B cells - that can be isolated and then immediately expanded using ImmunoCult™ Human B Cell Expansion Kit.
NB: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').