ImmunoCult™-XF B Cell Base Medium
Leverandør: STEMCELL Technologies
100-0646
STMC100-0646EA 0 NOK
STMC100-0646
ImmunoCult™-XF B Cell Base Medium
Media, Cellekultur
Serum-free and xeno-free base medium for the expansion of B cells.
Support the expansion of human B cells using serum- and xeno-free ImmunoCult™-XF B cell base medium. Available either individually or as part of ImmunoCult™ Human B cell expansion kit, this base medium is intended to be used in combination with ImmunoCult™-ACF Human B cell expansion supplement to prepare a complete serum-free medium for culturing and expanding human B cells.
NB: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').