NeuroCult™ SM1 Medium without Vitamin A
Leverandør: STEMCELL Technologies
100-1282
STMC100-1282EA 0 NOK
STMC100-1282
NeuroCult™ SM1 Medium without Vitamin A
Media, Cellekultur
Supplement (50X) without vitamin A for the culture of neural progenitor cells.
NeuroCult™ SM1 Without Vitamin A is a serum-free culture supplement, based on the published B27 formulation, with vitamin A removed. Vitamin A, also known as retinol, induces the differentiation of neural stem cells. In combination with a basal medium of choice, NeuroCult™ SM1 Without Vitamin A can be used for primary and pluripotent stem cell (PSC)-derived neural progenitor cell expansion and in customized differentiation protocols where vitamin A is not desired.
NB: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').