Anti-Perforin Mouse Monoclonal Antibody [clone: deltaG9]
Leverandør: STEMCELL Technologies
100-1457
STMC100-1457EA 0 NOK
STMC100-1457
Anti-Perforin Mouse Monoclonal Antibody [clone: deltaG9]
Antistoffer
Mouse monoclonal antibody against human perforin, unconjugated.
  • Nk cells
  • T cells CD8
This mouse monoclonal antibody (clone deltaG9) reacts with human perforin. Perforin is an ~70 kDa protein consisting of four domains: the N- and C-termini, which are related to its function, and two central domains that share similarities to complement proteins C6 - C9. Perforin is involved in the perforin/granzyme apoptosis pathway and is produced by natural killer and cytotoxic T cells. Upon contact with a target cell, perforin monomers are released and aggregate toMore Product Information