ImmunoCult™ Mouse Th2 Differentiation Supplement
Leverandør: STEMCELL Technologies
10955
STMC10955EA 0 NOK
STMC10955
ImmunoCult™ Mouse Th2 Differentiation Supplement
Media, Cellekultur
Supplement for the differentiation of mouse naïve CD4+ T cells into Th2 cells.
ImmunoCult™ Mouse Th2 Differentiation Supplement contains a combination of recombinant mouse cytokines and a blocking monoclonal antibody optimised to promote the differentiation of mouse naïve CD4+ T cells into Th2 cells. This supplement is intended for use with RPMI 1640 Medium containing fetal bovine serum and other additives, as well as anti-mouse CD3 and anti-mouse CD28 monoclonal antibodies as activating agents.
NB: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').