ImmunoCult™ Human Th1 Differentiation Supplement
Leverandør: STEMCELL Technologies
10973
STMC10973EA 0 NOK
STMC10973
ImmunoCult™ Human Th1 Differentiation Supplement
Media, Cellekultur
Culture supplement for the differentiation of human naïve CD4+ T cells into Th1 cells.
ImmunoCult™ Human Th1 Differentiation Supplement contains a combination of human cytokines and a blocking antibody formulated to promote the robust activation, expansion, and differentiation of peripheral blood-derived, naïve, CD4+ human T cells into Th1 cells. ImmunoCult™ Human Th1 Differentiation Supplement is intended for use with ImmunoCult™-XF T Cell Expansion Medium and ImmunoCult™ Human CD3/CD28 T Cell Activator.
This product is recommended for:
Research into the regulation of human naïve T cell differentiation and Th1 function
Development of procedures to expand and polarise naïve T cells toward a Th1 subset
NB: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').