RosetteSep™ Human Custom Cocktail
Leverandør: STEMCELL Technologies
15309
STMC15309EA 0 NOK
STMC15309
RosetteSep™ Human Custom Cocktail
Media, Celleseparasjon
Immunodensity isolation of untouched human cells.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Untouched, viable cells
  • Can be combined with SepMate™ for consistent, high-throughput sample processing
The RosetteSep™ Human Custom Cocktail is designed to deplete unwanted cells from fresh whole blood by negative selection. When centrifuged over a buoyant density medium such as Lymphoprep™ for more information.
NB: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including asMore Product Information