RosetteSep™ DM-M Density Medium
Leverandør: STEMCELL Technologies
15725
STMC15725EA 0 NOK
STMC15725
RosetteSep™ DM-M Density Medium
Media, Celleseparasjon
Density gradient medium for use with RosetteSep™ myeloid cell enrichment cocktail.
  • RosetteSep™ DM-M has a density of 1,085 g/ml
  • For use in cell isolation, In Vitro diagnostic applications
RosetteSep™ DM-M is a density gradient medium designed specifically for use with RosetteSep™ cocktails for the enrichment of specific human myeloid (CD33+) cells from whole blood.
NB: In light of the new EU Regulation 2017/746 (IVDR) of the European Parliament and of the Council on in vitro diagnostic medical devices, which is expected to become effective in 2022,More Product Information