SpinSep™ Density Medium
Leverandør: STEMCELL Technologies
17531
STMC17531EA 0 NOK
STMC17531
SpinSep™ Density Medium
Media, Celleseparasjon
Density gradient medium.
  • For use in cell isolation applications
SpinSep™ Density medium is a density gradient medium designed for use with the EasySep™ human myeloid positive selection kit.
NB: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’sMore Product Information