EasySep™ Human Naïve CD4+ T Cell Isolation Kit II
Leverandør: STEMCELL Technologies
17555 17555RF
STMC17555EA 0 NOK
STMC17555 STMC17555RF
EasySep™ Human Naïve CD4+ T Cell Isolation Kit II
Media, Celleseparasjon
Immunomagnetic isolation of untouched human naïve CD4+ T cells.
  • Fast, easy-to-use and column-free
  • Up to 98% purity
  • Untouched, viable cells
The EasySep™ Human Naïve CD4+ T Cell Isolation Kit II is designed to isolate naïve CD4+ T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring orMore Product Information