EasySep™ Direct Human CD8+ T Cell Isolation Kit
Leverandør: STEMCELL Technologies
19663
STMC19663EA 0 NOK
STMC19663
EasySep™ Direct Human CD8+ T Cell Isolation Kit
Media, Celleseparasjon
Immunomagnetic isolation of CD8+ T cells directly from whole blood
  • >99,9% red blood cell (RBC) depletion without the need for density gradient centrifugation, sedimentation, or lysis
  • Up to 91% purity of isolated cells
  • Fast, easy-to-use and column-free
  • Isolated cells are untouched
The EasySep™ Direct Human CD8+ T Cell Isolation Kit is designed to isolate CD8+ T cells from human whole blood by immunomagnetic negative selection. The EasySep™ Direct procedure involves labeling RBCs, platelets and unwanted cells present in human whole blood withMore Product Information