EasySep™ Red Blood Cell Lysis Buffer

Leverandør: STEMCELL Technologies

20120 20110
STMC20120EA 0 NOK
STMC20120 STMC20110
EasySep™ Red Blood Cell Lysis Buffer
Buffere Buffere, ekstrasjons

EasySep™ Red blood cell lysis buffer is for use as directed with EasySep™ or RoboSep™ whole blood kits for the lysis of red blood cells prior to separation from whole blood.


Application: Chimerism, Hla, immunology
Species: Human


NB: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR