EasySep™ Buffer

Leverandør: STEMCELL Technologies
20144
STMC20144EA 0 NOK
STMC20144
EasySep™ Buffer
Media, Celleseparasjon
EasySep™ buffer is recommended for EasySep™ cell separation protocols.

Application: Chimerism, Hla, immunology
Species: Human, mouse, non human primate, other, rat

NB: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use theMore Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR