RoboSep™ Buffer 2

Supplier: STEMCELL Technologies

20164
STMC20164EA 0 NOK
STMC20164
RoboSep™ Buffer 2
Buffers Biological Buffers

RoboSep™ buffer 2 is recommended for select EasySep™ cell separation protocols.


Application: Immunology
Species: Human, mouse, non human primate, other, rat


Please note that one or two bottles of buffer are included with every purchase of a RoboSep™ reagent kit.


Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR