Anti-CD138 Mouse Monoclonal Antibody [clone: MI15] (FITC)
Leverandør: STEMCELL Technologies
60003FI.1
STMC60003FI.1EA 0 NOK
STMC60003FI.1
Anti-CD138 Mouse Monoclonal Antibody [clone: MI15] (FITC)
Antistoffer
Mouse monoclonal IgG1 antibody against human, rhesus, cynomolgus CD138 (syndecan-1), FITC-conjugated.
  • B cells
  • Plasma
The MI15 antibody reacts with an extracellular epitope on CD138 (Syndecan-1), an ~85 - 95 kDa type 1 transmembrane glycoprotein expressed on the surface of pre-B cells, immature B cells, and normal and malignant plasma cells (but not mature circulating B cells), as well as on non-hematopoietic cells such as embryonic mesenchymal cells, endothelial, epithelial and neural cells. CD138 expression is used as a diagnostic marker for severalMore Product Information