Anti-CD14 Mouse Monoclonal Antibody [clone: M5E2] (PE)
Leverandør: STEMCELL Technologies
60004PE 60004PE.1
STMC60004PEEA 0 NOK
STMC60004PE STMC60004PE.1
Anti-CD14 Mouse Monoclonal Antibody [clone: M5E2] (PE)
Antistoffer
Mouse monoclonal IgG2a antibody against human, rhesus, cynomolgus CD14, PE-conjugated.
  • Monocytes
The M5E2 antibody reacts with CD14, an ~53 - 55 kDa GPI-anchored transmembrane glycoprotein expressed at high levels on the surface of peripheral blood monocytes and macrophages, and at lower levels on granulocytes. An ~10-fold difference in expression levels between monocytes/macrophages and granulocytes makes CD14 a useful marker for distinguishing these cell populations. CD14 is also found on tissue macrophages, Langerhans cells and dendritic cells. CD14More Product Information