Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10]
Leverandør: STEMCELL Technologies
60007
STMC60007EA 0 NOK
STMC60007
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10]
Antistoffer
Mouse monoclonal IgG1 antibody against human, rhesus, cynomolgus CD2, unconjugated.
  • T cells
The RPA-2.10 antibody reacts with CD2, an ~50 kDa type I transmembrane glycoprotein and a member of the immunoglobulin (Ig) superfamily; two Ig-like domains are located in its extracellular portion. CD2 is broadly expressed on peripheral T and NK cells, dendritic cells, erythrocytes, most thymic T cells, subsets of thymic B cells, and on the endothelium. CD2 expression appears early during T cell differentiation. Aberrant expression has been observed in someMore Product Information