Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (PE)

Leverandør: STEMCELL Technologies
60007PE 60007PE.1 100-0431
STMC60007PEEA 0 NOK
STMC60007PE STMC60007PE.1 STMC100-0431
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (PE)
Antistoffer
Mouse monoclonal IgG1 antibody against human, rhesus, cynomolgus CD2, PE-conjugated.

  • T cells

The RPA-2.10 antibody reacts with CD2, an ~50 kDa type I transmembrane glycoprotein and a member of the immunoglobulin (Ig) superfamily; two Ig-like domains are located in its extracellular portion. CD2 is broadly expressed on peripheral T and NK cells, dendritic cells, erythrocytes, most thymic T cells, subsets of thymic B cells, and on the endothelium. CD2 expression appears early during T cell differentiation. Aberrant expression has been observed in someMore Product Information
Bestill nå

SPESIFIKASJONER

Antigen-navn CD2
Environmentally Preferable
Klonalitet Monoclonal
Konjugering PE (Phycoerythrin)
Klon RPA-2.10
Reaktivitet Baboon,Capuchin monkey,Pig,Cynomolgus monkey,Rhesus monkey,Pigtailed macaque,Chimpanzee,Human,Squirrel monkey
Vert Mouse
Gen-ID 914
Isotype IgG1 kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR