Anti-CD25 Mouse Monoclonal Antibody [clone: HIT2]

Leverandør: STEMCELL Technologies
60014
STMC60014EA 0 NOK
STMC60014
Anti-CD25 Mouse Monoclonal Antibody [clone: HIT2]
Antistoffer
Mouse monoclonal IgG1 antibody against human, chimpanzee, horse CD38, unconjugated.

  • B cells
  • Hematopoietic stem
  • Progenitor cells
  • Plasma

The HIT2 antibody reacts with CD38, an ~45 kDa type II transmembrane glycoprotein expressed by many cell types, especially leukocytes. CD38 is an ectoenzyme (ADP-ribosyl hydrolase) with both cyclase and hydrolase activities, whereby it mediates lymphocyte activation, adhesion and metabolism. CD31 is a known ligand for the receptor. In humans, CD38 is expressed by the majority of hematopoietic cells at levels which varyMore Product Information
Bestill nå

SPESIFIKASJONER

Antigen-navn CD38
Environmentally Preferable
Klonalitet Monoclonal
Konjugering Unconjugated
Klon HIT2
Reaktivitet Horse,Chimpanzee,Human
Vert Mouse
Gen-ID 952
Isotype IgG1 kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR