Anti-CD4 Mouse Monoclonal Antibody [clone: OKT4] (PerCP-Cy5.5)

Leverandør: STEMCELL Technologies
60016PS 60016PS.1
STMC60016PSEA 0 NOK
STMC60016PS STMC60016PS.1
Anti-CD4 Mouse Monoclonal Antibody [clone: OKT4] (PerCP-Cy5.5)
Antistoffer
Mouse monoclonal IgG2b antibody against human, rhesus, cynomolgus CD4, PerCP-Cy5.5-conjugated.

  • T cells
  • T cells CD4
  • T cells Regulatory

The OKT4 antibody reacts with CD4, an ~59 kDa single-chain type 1 transmembrane glycoprotein and member of the immunoglobulin (Ig) superfamily; CD4 contains four extracellular Ig-like domains (D1 - D4). The epitope for OKT4 has been localised to the D3 domain of the protein, which has a structure resembling an Ig variable domain. CD4 is expressed at relatively high levels by most thymocytes and a subpopulation of TMore Product Information
Bestill nå

SPESIFIKASJONER

Antigen-navn CD4
Environmentally Preferable
Klonalitet Monoclonal
Konjugering PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
Klon OKT4
Reaktivitet Cynomolgus monkey,Sooty mangabey,Rhesus monkey,Chimpanzee,Human
Vert Mouse
Gen-ID 920
Isotype IgG2b kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR