Anti-CD49b Rat Monoclonal Antibody [clone: DX5]
Leverandør: STEMCELL Technologies
60020
STMC60020EA 0 NOK
STMC60020
Anti-CD49b Rat Monoclonal Antibody [clone: DX5]
Antistoffer
Rat monoclonal IgM antibody against mouse CD49b (integrin α2), unconjugated.
  • Nk cells
The DX5 antibody reacts with murine CD49b (integrin α2), an ~150 kDa type 1 transmembrane glycoprotein that associates non-covalently with CD29 (integrin β1) to form the heterodimeric CD49b/CD29 complex known as VLA-2, a receptor for extracellular matrix proteins such as collagen, E-cadherin, fibronectin, and laminin. CD49b is highly expressed by platelets and is found on a majority of NK cells, on NKT cells, and on a small subset of CD8+ T cells; the latterMore Product Information