Anti-CD45 Rat Monoclonal Antibody [clone: 30-F11] (PE)
Leverandør: STEMCELL Technologies
60030PE 60030PE.1
STMC60030PEEA 0 NOK
STMC60030PE STMC60030PE.1
Anti-CD45 Rat Monoclonal Antibody [clone: 30-F11] (PE)
Antistoffer
Rat monoclonal IgG2b antibody against mouse CD45, PE-conjugated.
  • Granulocytes
  • Subsets
  • Hematopoietic stem
  • Progenitor cells
  • Lymphocytes
  • Myeloid cells
The 30-F11 antibody reacts with an extracellular epitope found on all isoforms and both alloantigens (CD45.1 and CD45.2) of mouse CD45, a type I transmembrane glycoprotein expressed on the surface of most hematopoietic cells except mature erythrocytes, platelets and plasma cells; expression of CD45 is lost during differentiation of these cell types. CD45 is a member of the protein tyrosine phosphataseMore Product Information