Anti-CD138 Rat Monoclonal Antibody [clone: 281-2] (APC)
Leverandør: STEMCELL Technologies
60035AZ 60035AZ.1
STMC60035AZEA 0 NOK
STMC60035AZ STMC60035AZ.1
Anti-CD138 Rat Monoclonal Antibody [clone: 281-2] (APC)
Antistoffer
Rat monoclonal IgG2a antibody against mouse CD138 (syndecan-1), APC-conjugated.
  • B cells
  • Plasma
The 281-2 antibody reacts with an extracellular epitope on the core protein of mouse CD138 (Syndecan-1), a ~60 to 100 kDa type 1 transmembrane glycoprotein expressed on the surface of pre-B cells, immature B cells, and normal and malignant plasma cells (but not mature circulating B cells). CD138 is also expressed on non-hematopoietic cells such as embryonic mesenchymal cells, endothelial, epithelial and neural cells. CD138 is thought to act primarily as aMore Product Information