Anti-CD105 Mouse Monoclonal Antibody [clone: 43A3] (FITC)

Leverandør: STEMCELL Technologies
60039FI 60039FI.1
STMC60039FIEA 0 NOK
STMC60039FI STMC60039FI.1
Anti-CD105 Mouse Monoclonal Antibody [clone: 43A3] (FITC)
Antistoffer
Mouse monoclonal IgG1 antibody against human, mouse CD105 (endoglin), FITC-conjugated.

  • Hematopoietic stem
  • Progenitor cells
  • Mesenchymal stem
  • Progenitor cells

The 43A3 antibody reacts with CD105 (endoglin), an ~180 kDa cell surface glycoprotein which is a disulfide-bonded homodimer of ~90 kDa type I transmembrane subunits. CD105 is a component of the TGF-β receptor complex and is expressed by vascular endothelial smooth muscle cells, syncytiotrophoblasts of placenta and activated macrophages, and at relatively low levels by stromal fibroblasts. ItsMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol CD105
Antigen-navn Endoglin
Environmentally Preferable
Klonalitet Monoclonal
Konjugering FITC (Fluorescein Isothiocyanate)
Klon 43A3
Reaktivitet Human,Mouse
Vert Mouse
Gen-ID 2022
Isotype IgG1 kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR