Anti-CD73 Mouse Monoclonal Antibody [clone: AD2] (FITC)

Leverandør: STEMCELL Technologies
60044FI 60044FI.1
STMC60044FIEA 0 NOK
STMC60044FI STMC60044FI.1
Anti-CD73 Mouse Monoclonal Antibody [clone: AD2] (FITC)
Antistoffer
Mouse monoclonal IgG1 antibody against human, rhesus, chimpanzee CD73, FITC-conjugated.

  • Mesenchymal stem
  • Progenitor cells

The AD2 antibody reacts with human CD73, a glycosyl phosphatidylinositol (GPI)-anchored glycoprotein and ecto-5’-nucleotidase expressed on the surface of subsets of B and T cells, follicular dendritic cells, mesenchymal stem cells, endothelial cells, and epithelial cells. CD73 catalyses the hydrolysis of 5’-adenosine monophosphate (AMP) to form the bioactive nucleoside adenosine, and plays a pivotal role in the activation of P1More Product Information
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn CD73
Klonalitet Monoclonal
Konjugering FITC (Fluorescein Isothiocyanate)
Klon AD2
Reaktivitet Rhesus monkey,Macaque,Chimpanzee,Human
Vert Mouse
Gen-ID 4907
Isotype IgG1 kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR