Anti-CD73 Mouse Monoclonal Antibody [clone: AD2] (PE)
Leverandør: STEMCELL Technologies
60044PE 60044PE.1
STMC60044PEEA 0 NOK
STMC60044PE STMC60044PE.1
Anti-CD73 Mouse Monoclonal Antibody [clone: AD2] (PE)
Antistoffer
Mouse monoclonal IgG1 antibody against human, rhesus, chimpanzee CD73, PE-conjugated.
  • Mesenchymal Stem
  • Progenitor cells
The AD2 antibody reacts with human CD73, a glycosyl phosphatidylinositol (GPI)-anchored glycoprotein and ecto-5’-nucleotidase expressed on the surface of subsets of B and T cells, follicular dendritic cells, mesenchymal stem cells, endothelial cells, and epithelial cells. CD73 catalyzes the hydrolysis of 5’-adenosine monophosphate (AMP) to form the bioactive nucleoside adenosine, and plays a pivotal role in the activation of P1More Product Information