Anti-SSEA-4 Mouse Monoclonal Antibody [clone: MC-813-70] (FITC)

Leverandør: STEMCELL Technologies
60062FI 60062FI.1
STMC60062FIEA 0 NOK
STMC60062FI STMC60062FI.1
Anti-SSEA-4 Mouse Monoclonal Antibody [clone: MC-813-70] (FITC)
Antistoffer
Mouse monoclonal IgG3 antibody against human, mouse, rat SSEA-4, FITC-conjugated.

  • Pluripotent Stem cells

The MC-813-70 antibody reacts with stage-specific embryonic antigen-4 (SSEA-4), a glycolipid carbohydrate antigen expressed on the surface of human embryonal carcinoma (EC), embryonic germ (EG), undifferentiated embryonic stem (ES) and induced pluripotent stem (iPS) cells, a subset of mesenchymal stem cells, and rhesus monkey ES cell lines. No immunoreactivity is evident with undifferentiated mouse EC, EG, ES, and iPS cells. Expression of SSEA-4 isMore Product Information
Bestill nå

SPESIFIKASJONER

Antigen-navn SSEA-4
Environmentally Preferable
Klonalitet Monoclonal
Konjugering FITC (Fluorescein Isothiocyanate)
Klon MC-813-70
Reaktivitet Rabbit,Rhesus monkey,Chicken,Human,Dog,Rat,Mouse,Cat
Vert Mouse
Gen-ID 330401
Isotype IgG3 kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR