Anti-Igg1 Isotype Control Rat Monoclonal Antibody [clone: RTK2071] (Biotin)
Leverandør: STEMCELL Technologies
60075BT 60075BT.1
STMC60075BTEA 0 NOK
STMC60075BT STMC60075BT.1
Anti-Igg1 Isotype Control Rat Monoclonal Antibody [clone: RTK2071] (Biotin)
Antistoffer
Rat monoclonal IgG1, kappa isotype control antibody, biotin-conjugated.
The RTK2071 antibody (IgG1, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody. Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used. If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen. The use of an appropriateMore Product Information