Anti-Igg2b Isotype Control Rat Monoclonal Antibody [clone: RTK4530] (Alexa Fluor® 488)
Leverandør: STEMCELL Technologies
60077AD
STMC60077ADEA 0 NOK
STMC60077AD
Anti-Igg2b Isotype Control Rat Monoclonal Antibody [clone: RTK4530] (Alexa Fluor® 488)
Antistoffer
Rat monoclonal IgG2b, kappa isotype control antibody, Alexa Fluor® 488-conjugated.
The RTK4530 antibody (IgG2b, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody. Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used. If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen. The use of anMore Product Information