Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (Biotin)
Leverandør: STEMCELL Technologies
60082BT
STMC60082BTEA 0 NOK
STMC60082BT
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (Biotin)
Antistoffer
Mouse monoclonal IgG1 antibody against human, rhesus, cynomolgus CD5, biotin-conjugated.
  • T cells
The UCHT2 antibody reacts with CD5, an ~67 kDa single-chain type I glycoprotein and member of the scavenger receptor superfamily, which is constitutively expressed on thymocytes, T cells, B cell subsets, peripheral blood dendritic cells and some leukemia and lymphoma cells, including chronic B lymphocytic leukemia (B-CLL) cells. CD5 is expressed at low levels on thymocytes and at high density on mature T cells. Putative ligands include CD5L and CD72. CD5More Product Information
Bestill nå
SPESIFIKASJONER
Antigen-navn CD5
Environmentally Preferable
Klonalitet Monoclonal
Konjugering Biotin
Klon UCHT2
Reaktivitet Capuchin monkey,Cynomolgus monkey,Common marmoset,Rhesus monkey,Chimpanzee,Human,Owl monkey
Vert Mouse
Gen-ID 921
Isotype IgG1 kappa