Anti-c-KIT Mouse Monoclonal Antibody [clone: 104D2] (APC)

Leverandør: STEMCELL Technologies
60087AZ 60087AZ.1
STMC60087AZEA 0 NOK
STMC60087AZ STMC60087AZ.1
Anti-c-KIT Mouse Monoclonal Antibody [clone: 104D2] (APC)
Antistoffer
Mouse monoclonal IgG1 antibody against human, cynomolgus, cow CD117 (c-Kit), APC-conjugated.

  • Endoderm Psc Derived
  • Hematopoietic stem
  • Progenitor cells

The 104D2 antibody reacts with CD117 (c-kit), an ~145 kDa type 1 transmembrane receptor for c-kit ligand (stem cell factor/steel factor) that is broadly expressed on hematopoietic stem cells in bone marrow, including pluripotent and erythroid progenitor cells and B and T lymphocyte precursors, as well as on mast cells and melanocytes. CD117 is expressed on ~1 to 4% of bone marrow cells, the majority ofMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol CD117
Antigen-navn c-KIT
Environmentally Preferable
Klonalitet Monoclonal
Konjugering APC (Allophycocyanin)
Klon 104D2
Reaktivitet Cynomolgus monkey,Human,Bovine
Vert Mouse
Gen-ID 3815
Isotype IgG1 kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR