Anti-CD117 Mouse Monoclonal Antibody [clone: 104D2] (PE)
Leverandør: STEMCELL Technologies
60087PE 60087PE.1
STMC60087PEEA 0 NOK
STMC60087PE STMC60087PE.1
Anti-CD117 Mouse Monoclonal Antibody [clone: 104D2] (PE)
Antistoffer
Mouse monoclonal IgG1 antibody against human, cynomolgus, cow CD117 (c-Kit), PE-conjugated.
  • Endoderm Psc derived
  • Hematopoietic stem
  • Progenitor cells
The 104D2 antibody reacts with CD117 (c-Kit), an ~145 kDa type 1 transmembrane receptor for c-Kit ligand (stem cell factor/steel factor) that is broadly expressed on hematopoietic stem cells in bone marrow, including pluripotent and erythroid progenitor cells and B and T lymphocyte precursors, as well as on mast cells and melanocytes. CD117 is expressed on ~1 - 4% of bone marrow cells, the majority ofMore Product Information