Anti-CD45RO Mouse Monoclonal Antibody [clone: UCHL1] (APC)

Leverandør: STEMCELL Technologies
60097AZ 60097AZ.1
STMC60097AZEA 0 NOK
STMC60097AZ STMC60097AZ.1
Anti-CD45RO Mouse Monoclonal Antibody [clone: UCHL1] (APC)
Antistoffer
Mouse monoclonal IgG2a antibody against human, chimpanzee, common marmoset CD45RO, APC-conjugated.

  • Leukocytes
  • T cells
  • T cells CD4
  • T cells CD8

The UCHL1 antibody reacts with an extracellular epitope on CD45RO, which is the shortest isoform of CD45, a type I transmembrane glycoprotein and member of the protein tyrosine phosphatase family (receptor class 1/6 subfamily). Alternative splicing of exons 4, 5 and 6 that encode the extracellular RA, RB and RC polypeptides of CD45 gives rise to up to 8 isoforms with molecular masses of 180 - 240 kDa. ExcisionMore Product Information
Bestill nå

SPESIFIKASJONER

Antigen-navn CD45RO
Environmentally Preferable
Klonalitet Monoclonal
Konjugering APC (Allophycocyanin)
Klon UCHL1
Reaktivitet Common marmoset,Chimpanzee,Human
Vert Mouse
Gen-ID 5788
Isotype IgG2a kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR