Anti-CD71 Mouse Monoclonal Antibody [clone: OKT9] (Biotinilated)
Leverandør: STEMCELL Technologies
60106BT
STMC60106BTEA 0 NOK
STMC60106BT
Anti-CD71 Mouse Monoclonal Antibody [clone: OKT9] (Biotinilated)
Antistoffer
Mouse monoclonal IgG1 antibody against human CD71 (transferrin receptor), biotin-conjugated.
  • Erythroid cells
The OKT9 antibody reacts with CD71 (transferrin receptor), an ~180 - 190 kDa disulfide-bonded homodimer and type 2 transmembrane glycoprotein expressed at high levels on a broad range of actively proliferating cells, as well as reticulocytes, monocytes, macrophages and marrow stromal cells. Surface levels are low on resting leukocytes but are upregulated on lymphocytes, monocytes and macrophages following antigen or mitogen stimulation.More Product Information