Anti-CD83 Mouse Monoclonal Antibody [clone: HB15e] (FITC)

Leverandør: STEMCELL Technologies
60107FI 60107FI.1
STMC60107FIEA 0 NOK
STMC60107FI STMC60107FI.1
Anti-CD83 Mouse Monoclonal Antibody [clone: HB15e] (FITC)
Antistoffer
Mouse monoclonal IgG1 antibody against human, rhesus, cynomolgus CD83, FITC-conjugated.

  • Dendritic cells

The HB15e antibody reacts with CD83, an ~45 kDa type I transmembrane glycoprotein and member of the Siglec protein family, containing an extracellular V-type Ig-like domain and a C-terminal cytoplasmic tail. A soluble form of CD83 has also been identified. CD83 is expressed predominantly on circulating and tissue dendritic cells, including Langerhan cells and interdigitating reticulum cells. It is expressed weakly on activated lymphocytes and mayMore Product Information
Bestill nå

SPESIFIKASJONER

Antigen-navn CD83
Environmentally Preferable
Klonalitet Monoclonal
Konjugering FITC (Fluorescein Isothiocyanate)
Klon HB15e
Reaktivitet Baboon,Cynomolgus monkey,Rhesus monkey,Macaque,Chimpanzee,Human
Vert Mouse
Gen-ID 9308
Isotype IgG1 kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR