Anti-Igg2a Isotype Control Mouse Monoclonal Antibody [clone: C1.18.4] (PE)

Leverandør: STEMCELL Technologies
60108PE
STMC60108PEEA 0 NOK
STMC60108PE
Anti-Igg2a Isotype Control Mouse Monoclonal Antibody [clone: C1.18.4] (PE)
Antistoffer
Mouse monoclonal IgG2a, kappa isotype control antibody, PE-conjugated.

The C1.18.4 antibody (IgG2a, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody. Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being employed. If a conjugated antibody is employed, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen. The use of anMore Product Information
Bestill nå

SPESIFIKASJONER

Antigen-navn Igg2a Isotype Control
Environmentally Preferable
Klonalitet Monoclonal
Konjugering PE (Phycoerythrin)
Klon C1.18.4
Reaktivitet
Vert Mouse
Isotype IgG2a kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR