Anti-CD19 Rat Monoclonal Antibody [clone: 1D3] (PerCP-Cy5.5)

Leverandør: STEMCELL Technologies
60112PS 60112PS.1
STMC60112PSEA 0 NOK
STMC60112PS STMC60112PS.1
Anti-CD19 Rat Monoclonal Antibody [clone: 1D3] (PerCP-Cy5.5)
Antistoffer
Rat monoclonal IgG2a antibody against mouse CD19, PerCP-Cy5.5-conjugated.

  • B cells

The 1D3 antibody reacts with mouse CD19, an ~95 kDa type 1 transmembrane glycoprotein expressed on the surface of B cells throughout all stages of development, from early pre-B cells to plasma cells. Expression is down-regulated but persists in terminally differentiated plasma cells. CD19 is also found on follicular dendritic cells. By associating with CD21 and CD81, CD19 functions as a co-receptor for the B cell receptor and is involved in B cell activation andMore Product Information
Bestill nå

SPESIFIKASJONER

Antigen-navn CD19
Environmentally Preferable
Klonalitet Monoclonal
Konjugering PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Klon 1D3
Reaktivitet Mouse
Vert Rat
Gen-ID 12478
Isotype IgG2a kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR