Anti-CD41 Mouse Monoclonal Antibody [clone: HIP8] (PE)

Leverandør: STEMCELL Technologies
60114PE 60114PE.1
STMC60114PEEA 0 NOK
STMC60114PE STMC60114PE.1
Anti-CD41 Mouse Monoclonal Antibody [clone: HIP8] (PE)
Antistoffer
Mouse monoclonal IgG1 antibody against human, rhesus, cynomolgus CD41, PE-conjugated.

  • Megakaryocytes
  • Platelets

The HIP8 antibody reacts with the ~125 kDa GPIIbα subunit of human CD41, a heterodimeric integrin expressed primarily on platelets, megakaryocytes, monocytes, and acute megakaryoblastic leukemia cells. The GPIIbα subunit is linked to a smaller ~25 kDa subunit (GPIIbβ) by a single disulfide bond. Three isoforms of CD41 that are generated by alternative splicing have been identified, one of which is expressed only by cancerous cells. CD41More Product Information
Bestill nå

SPESIFIKASJONER

Antigen-navn CD41
Environmentally Preferable
Klonalitet Monoclonal
Konjugering PE (Phycoerythrin)
Klon HIP8
Reaktivitet Baboon,Capuchin monkey,Pig,Cynomolgus monkey,Rhesus monkey,Chimpanzee,Human
Vert Mouse
Gen-ID 3674
Isotype IgG1 kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR