Anti-CD34 Mouse Monoclonal Antibody [clone: 8G12] (PE)
Leverandør: STEMCELL Technologies
60121PE
STMC60121PEEA 0 NOK
STMC60121PE
Anti-CD34 Mouse Monoclonal Antibody [clone: 8G12] (PE)
Antistoffer
Mouse monoclonal IgG1 antibody against human CD34, PE-conjugated.
  • Hematopoietic stem
  • Progenitor cells
CD34 is an ~105 - 120 kDa type 1 transmembrane glycoprotein expressed on the surface of most human hematopoietic stem and progenitor cells (HSPCs), as well as on endothelial cells and some tumor cell lines. The frequency of CD34+ cells is low in bone marrow (1 - 4%), cord blood (~1%), and peripheral blood (<0,1%). CD34 is used to identify and isolate HSPCs, including those capable of reconstituting hematopoiesis after transplantation. DistinctMore Product Information