Anti-E-cadherin (CD324) Rat Monoclonal Antibody [clone: DECMA-1]

Leverandør: STEMCELL Technologies
60157 60157.1
STMC60157EA 0 NOK
STMC60157 STMC60157.1
Anti-E-cadherin (CD324) Rat Monoclonal Antibody [clone: DECMA-1]
Antistoffer
Rat monoclonal IgG1 antibody against human, mouse, dog E-cadherin (CD324), unconjugated.

  • Airway cells

The DECMA-1 antibody recognizes E-cadherin, a calcium-dependent transmembrane glycoprotein involved in cell-cell adhesion, bacterial pathogenesis, and tumor invasion. The extracellular region of E-cadherin is composed of multiple cadherin repeats that contain calcium-binding motifs. The cytoplasmic domain of E-cadherin is reported to interact with a range of intracellular proteins including erbin, ezrin, caspase-3, caspase-8, β-catenin, presenilin 1,More Product Information
Bestill nå

SPESIFIKASJONER

Antigen-navn E-cadherin (CD324)
Environmentally Preferable
Klonalitet Monoclonal
Konjugering Unconjugated
Klon DECMA-1
Reaktivitet Pig,Human,Dog,Mouse
Vert Rat
Gen-ID 999/12550
Isotype IgG1 kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR