Anti-HLA-DR Mouse Monoclonal Antibody [clone: LN3]
Leverandør: STEMCELL Technologies
60164
STMC60164EA 0 NOK
STMC60164
Anti-HLA-DR Mouse Monoclonal Antibody [clone: LN3]
Antistoffer
Mouse monoclonal IgG2b antibody against human, rhesus HLA-DR, unconjugated.
  • B cells
  • Dendritic cells
  • Macrophages
  • Monocytes
  • Other
  • T cells
The LN3 antibody reacts with the HLA-DR antigen, a major histocompatibility complex (MHC) class II receptor encoded within the human leukocyte antigen (HLA) complex on chromosome 6. HLA-DR is a heterodimeric transmembrane glycoprotein comprising non-covalently associated α (36 kDa) and β (27 kDa) subunits. MHC class II plays a central role in the presentation of antigen-derived peptides to CD4+ T cells, along with CD3/More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Antigen-navn HLA-DR
Klonalitet Monoclonal
Konjugering Unconjugated
Klon LN3
Reaktivitet Rhesus monkey,Human
Vert Mouse
Gen-ID 3123
Isotype IgG2b kappa