Propidium iodide cell viability stain

Leverandør: STEMCELL Technologies

75002
STMC75002EA 0 NOK
STMC75002
Propidium iodide cell viability stain
Propidium iodide

Propidium Iodide (PI) is a red-fluorescent cell viability dye which is excluded from live cells with intact membranes, but penetrates dead or damaged cells and binds to DNA and RNA by intercalating between the bases.


  • Cell viability dye (DNA-labeling dye)


It is widely used as a counterstain to differentiate and exclude non-viable cells in flow cytometric analyses, and can be excited using blue (488 nm), green (532 nm), or yellow-green (561 nm) laser lines, with detection in the FL2 or FL3 channels. PI is used in DNA fluorescence imaging applications to discriminate early and late stages of apoptosis, to study cell‐mediated cytotoxicity, and for chromosome analysis. It is also commonly used in quantitative DNA assays.


NB: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Formel: C₂₇H₃₄I₂N₄
MDL nummer: MFCD00011921
CAS-nummer: 25535-16-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR