Resazurin sodium salt

Leverandør: STEMCELL Technologies

Synonymer: 7-Sodiooxy-3H-phenoxazin-3-one 10-oxide, Sodium 7-oxido-3-oxo-3H-phenoxazine 10-oxide, Resazurin sodium, Resazurin

75005
STMC75005EA 0 NOK
STMC75005
Resazurin sodium salt
Resazurin sodium salt

Resazurin (7-hydroxy-10-oxidophenoxazin-10-ium-3-one, sodium) is a blue fluorogenic dye used as a redox indicator in cell viability and proliferation assays for bacteria, yeast or mammalian cells.


  • Cell proliferation and viability dye


The blue form of the dye is irreversibly reduced by enzymes in viable cells to generate a highly red-fluorescent product, Resazurin, which exhibits an emission maximum at ~580 nm and can be detected by flow cytometry, fluorescence microscopy, and high-throughput screening methods. Resazurin is minimally toxic to living cells, making it suitable for use in long-term cell culture. The dye has also been used to assay L-glutamate and to measure the metabolic activity of mitochondria.


NB: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Formel: C₁₂H₆NNaO₄
MW: 251.17 g/mol
MDL nummer: MFCD00005036
CAS-nummer: 62758-13-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR