dNTP Mix (10 mM)
Leverandør: STEMCELL Technologies
76027
STMC76027EA 0 NOK
STMC76027
dNTP Mix (10 mM)
Transfeksjonsreagenser
A solution containing sodium salts of dATP, dCTP, dGTP and dTTP, each at 10 mM.
  • Ready-to-use
  • Animal component-free
  • High purity (>99% triphosphate)
Simplify a variety of molecular biology processes by using dNTP Mix, a solution that contains a mixture of four nucleotides (dATP, dCTP, dGTP, dTTP). Each nucleotide is at a concentration of 10 mM in water at pH 7,5 and has greater than 99% purity. This dNTP Mix can be used in applications such as polymerase chain reaction (PCR), sequencing, fill-in reactions, and cDNA synthesis.
NB: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').