Human Recombinant IL-1 beta
Leverandør: STEMCELL Technologies
78034 78034.1 78034.3
STMC78034EA 0 NOK
STMC78034 STMC78034.1 STMC78034.3
Human Recombinant IL-1 beta
Proteiner
Interleukin 1 beta (IL-1β) is synthesized as an inactive precursor protein or pro-IL-1β. This precursor is cleaved intracellularly by caspase 1 (IL-1β convertase) to form the active form of the protein that is later secreted (Allan et al.). IL-1β binds to IL-1 receptor and activates intracellular signaling via the MAPK or NF-κB pathway. IL-1β is released by monocytes, tissue macrophages, and dendritic cells in response to infection or injury and induces expression of acute-phase proteins. It also promotes the infiltration of inflammatory and immunocompetent cells from the circulation into the extravascular space and affected tissues, by stimulating the expression of adhesion molecules on endothelial cells. IL-1β also affects other immune cells; for example, it co-stimulates T cell functions together with an antigen or mitogen. It also stimulates Th17 differentiation and B cell proliferation in an IL-6-dependent manner.
NB: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Bestill nå
SPESIFIKASJONER
Protein/peptide name IL-1 beta
Environmentally Preferable
Protein/peptide type Recombinant
Arter Human
Kilde
Renhet ≥95%