STEMscript™ cDNA Synthesis Kit with Random Primers
Leverandør: STEMCELL Technologies
79004
STMC79004EA 0 NOK
STMC79004
STEMscript™ cDNA Synthesis Kit with Random Primers
Transfeksjonsreagenser
For reverse transcription of RNA without poly(A) tails, before qPCR or RNA sequencing.
Use STEMscript™ cDNA Synthesis Kit with Random Primers for reverse transcription of RNA without poly(A) tails (e.g. rRNA, tRNA, degraded RNA, RNA isolated from prokaryotic sources, and RNA with known secondary structures) prior to qPCR or RNA sequencing.
This kit includes three components that have been optimised to provide efficient synthesis of first-strand cDNA in preparation for qPCR amplification: STEMscript™ Reverse Transcriptase Mix (contains STEMscript™ Reverse Transcriptase purified from recombinant E. coli, and recombinant ribonuclease inhibitor), STEMscript™ Reaction Buffer (contains primers, MgCl₂, and dNTPs), and nuclease-free water.
Random primers are ideal for reverse transcription of RNA without poly(A) tails. For reverse transcription of eukaryotic mRNA with poly(A) tails, please use STEMscript™ cDNA Synthesis Kit with Oligo(dT) Primers, which includes primers that anneal to the poly(A) tails.
NB: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').