Cole-Parmer® SHP-200 Series Large Capacity Infrared Stirring Hotplate
Leverandør: Cole Parmer
04807-86
STUA04807-86EA 21225 NOK
STUA04807-86
Cole-Parmer® SHP-200 Series Large Capacity Infrared Stirring Hotplate
Omrørere Magnetrørere med varmeplate
Efficient infrared heater saves time and energy.
  • Large plate surface is ideal for heating large volumes of liquid
  • Ceramic top stays cool around the edges, preventing burns
  • Be safe - flashing light warns when top plate is hot
  • BioCote® antimicrobial protection is built in for a cleaner, more hygienic product
Efficient infrared heater combined with a stirrer delivers faster, evenly heated fluids. A 900-watt infrared heater can boil a litre of water over 30% faster than a conventional, 1200-watt ceramic hotplate. Rare earth magnets provide powerfulMore Product Information